accessibility menu, dialog, popup

UserName

HOONHNH2

CAS RN 13349-82-1

IUPAC Name: 4-[2-(2-hydroxyethoxy)ethyl]piperazine-1,4-diium
Synonyms: 4-[2-(2-hydroxyethoxy)ethyl]piperazine-1,4-diium
Molecular Weight (g/mol): 176.26
Molecular Formula: C8H20N2O2
InChi Key: FLNQAPQQAZVRDA-UHFFFAOYSA-P
SMILES: OCCOCC[NH+]1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)