accessibility menu, dialog, popup

UserName

H3COFF

CAS RN 134364-69-5

IUPAC Name: 1,2-difluoro-3-methoxybenzene
Synonyms: 1,2-difluoro-3-methoxybenzene
Molecular Weight (g/mol): 144.12
Molecular Formula: C7H6F2O
InChi Key: RDOGTTNFVLSBKG-UHFFFAOYSA-N
SMILES: COC1=CC=CC(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results