missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoroanisole, 97+%, Thermo Scientific™

Product Code. 11499967
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499967 5 g 5g
11489967 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499967 Supplier Thermo Scientific Alfa Aesar Supplier No. B22916.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134364-69-5
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD01631562
InChI Key RDOGTTNFVLSBKG-UHFFFAOYSA-N
Synonym 2,3-difluoroanisole, 1,2-difluoro-3-methoxy-benzene, 2,3-fluoroanisole, 2,3-difluoro-1-methoxybenzene, benzene, 1,2-difluoro-3-methoxy, 2,3-difluoroanisol, difluro-methoxybenzen, pubchem1969, 2,3-difluoro anisole, acmc-1bwhl
PubChem CID 2769344
IUPAC Name 1,2-difluoro-3-methoxybenzene
SMILES COC1=CC=CC(F)=C1F

Specifications

Flash Point 62°C (144°F)
Quantity 5 g
Formula Weight 144.12
Percent Purity ≥97%
Chemical Name or Material 2,3-Difluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.