CAS RN 1363281-27-9
CAS RN 1363281-27-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Tocris Bioscience™ VU 0409551
Selective mGlu5 positive allosteric modulator; brain penetrant and orally bioavailable
2
JNJ-46778212, MedChemExpress
MedChemExpress JNJ-46778212 (VU 0409551) is an mGlu5 positive allosteric modulator with an EC50 of 260 nM.
3
Tocris Bioscience™ VU 0409551
Selective mGlu5 positive allosteric modulator; brain penetrant and orally bioavailable
4
Tocris Bioscience™ VU 0409551
Selective mGlu5 positive allosteric modulator; brain penetrant and orally bioavailable