missing translation for 'onlineSavingsMsg'
Learn More

JNJ-46778212, MedChemExpress

Product Code. 16467808
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16467808

Brand: Medchem Express HY19559

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ-46778212 (VU 0409551) is an mGlu5 positive allosteric modulator with an EC50 of 260 nM.

JNJ-46778212 (VU 0409551) is an mGlu5 positive allosteric modulator with an EC50 of 260 nM.

Specifications

Chemical Name or Material JNJ-46778212
CAS 1363281-27-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H17FN2O3
Quantity 10 mg
Synonym VU 0409551
Solubility Information DMSO : ≥ 125 mg/mL (354.75 mM)
SMILES O=C(N1CCC2=C(OC(COC3=CC=CC=C3)=N2)C1)C4=CC=C(F)C=C4
Molecular Weight (g/mol) 352.36
Formula Weight 352.36
Percent Purity 99.46%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.