accessibility menu, dialog, popup

UserName

CH3OOOH3CNH2Br

CAS RN 1378874-22-6

IUPAC Name: methyl 2-amino-3-bromo-5-methoxybenzoate
Synonyms: methyl 2-amino-3-bromo-5-methoxybenzoate
Molecular Weight (g/mol): 260.09
Molecular Formula: C9H10BrNO3
InChi Key: WPRSGBOUSTUNEC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=CC(Br)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results