missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-amino-3-bromo-5-methoxybenzoate, 96%, Thermo Scientific Chemicals

Product Code. 15491008
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491008 1 g 1g
15401018 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491008 Supplier Thermo Scientific Alfa Aesar Supplier No. H54844.03
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1378874-22-6 | Molecular Formula: C9H10BrNO3 | Molecular Weight: 260.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1378874-22-6
Molecular Formula C9H10BrNO3
Molecular Weight (g/mol) 260.087
MDL Number MFCD20527215
InChI Key WPRSGBOUSTUNEC-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-methoxybenzoic acid methyl ester
PubChem CID 73995991
IUPAC Name methyl 2-amino-3-bromo-5-methoxybenzoate
SMILES COC1=CC(=C(C(=C1)C(=O)OC)N)Br

Specifications

Quantity 1 g
Formula Weight 260.09
Percent Purity 96%
Chemical Name or Material Methyl 2-amino-3-bromo-5-methoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.