CAS RN 13865-21-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-Methyl-4-oxobutanoate (Technical Grade), TRC
CAS: 13865-21-9 Molecular Formula: C6H10O3 Molecular Weight (g/mol): 130.14 IUPAC Name: methyl 2-methyl-4-oxobutanoate SMILES: COC(=O)C(C)CC=O