accessibility menu, dialog, popup

UserName

H3CNHNNHH3CCH3NS(R)HNCH3OOHON

CAS RN 138722-96-0

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Industry Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results