missing translation for 'onlineSavingsMsg'
Learn More

Atrazine Mercapturate, TRC

Product Code. 30133398
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133398 50 mg 50mg
30006469 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133398 Supplier Toronto Research Chemicals Supplier No. TRCA79461550MG
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138722-96-0
Molecular Formula C13 H22 N6 O3 S
Molecular Weight (g/mol) 342.42
Synonym N-Acetyl-S-[4-(ethylamino)-6-[(1-methylethyl)amino]-1,3,5-triazin-2-yl]-L-cysteine, Atrazine mercapturate, Atrazine-mercapturate
IUPAC Name (2R)-2-acetamido-3-[[4-(ethylamino)-6-(propan-2-ylamino)-1,3,5-triazin-2-yl]sulfanyl]propanoic acid
SMILES CCNc1nc(NC(C)C)nc(SC[C@H](NC(=O)C)C(=O)O)n1

Specifications

Analyte or Component Names Atrazine-mercapturate
Shipping Condition Room Temperature
InChI Formula InChI=1S/C13H22N6O3S/c1-5-14-11-17-12(15-7(2)3)19-13(18-11)23-6-9(10(21)22)16-8(4)20/h7,9H,5-6H2,1-4H3,(H,16,20)(H,21,22)(H2,14,15,17,18,19)/t9-/m0/s1
Solution Type Neat
Industry Type Environmental Testing
Quantity 50 mg
Formula Weight 342.1474
Chemical Name or Material Atrazine-mercapturate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.