accessibility menu, dialog, popup

UserName

P

CAS RN 13885-09-1

IUPAC Name: {[1,1'-biphenyl]-2-yl}diphenylphosphane
Synonyms: [1,1'-biphenyl]-2-yldiphenylphosphane
Molecular Weight (g/mol): 338.39
Molecular Formula: C24H19P
InChi Key: FNCQSSIMHQVKGF-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results