missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)biphenyl, 98%, Thermo Scientific Chemicals

Product Code. 15468198
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468198 5 g 5g
15478198 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468198 Supplier Thermo Scientific Alfa Aesar Supplier No. H61640.06
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

2-(Diphenylphosphino)biphenyl is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Diphenylphosphino)biphenyl is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13885-09-1
Molecular Formula C24H19P
Molecular Weight (g/mol) 338.39
MDL Number MFCD11559063
InChI Key FNCQSSIMHQVKGF-UHFFFAOYSA-N
Synonym 2-diphenylphosphino biphenyl, 2-diphenylphosphino-biphenyl, 1,1'-biphenyl-2-yldiphenylphosphane, biphenyl-2-yl-diphenyl-phosphane, phosphine, 1,1'-biphenyl yldiphenyl, diphenyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldiphenyl-phosphine, ksc494o1b, 2-biphenylyl diphenylphosphine, diphenylphosphino-1,1'-biphenyl
PubChem CID 12976978
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
IUPAC Name diphenyl-(2-phenylphenyl)phosphane
Formula Weight 338.38
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-(Diphenylphosphino)biphenyl

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.