accessibility menu, dialog, popup

UserName

CAS RN 139-02-6

IUPAC Name: sodium benzenolate
Synonyms: sodium phenoxide
Molecular Weight (g/mol): 116.1
Molecular Formula: C6H5NaO
InChi Key: NESLWCLHZZISNB-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)