missing translation for 'onlineSavingsMsg'
Learn More

Sodium phenoxide, 98%

Product Code. 11431597
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11431597 50 g 50g
11441597 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11431597 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor B25002.18
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 139-02-6 | C6H5NaO | 116.10 g/mol

Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phenoxide is used as a precursor to aryl ethers and involved in the preparation of phenyl ethers and metal phenolates. It serves as an antiseptic and used in organic synthesis.

Solubility
Very soluble in water, soluble in alcoholSoluble in water, acetone and alcohol.

Notes
Hygroscopic and air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 139-02-6
Molecular Formula C6H5NaO
Molecular Weight (g/mol) 116.10
MDL Number MFCD00013134
InChI Key NESLWCLHZZISNB-UHFFFAOYSA-M
Synonym sodium phenoxide, sodium phenolate, sodium phenate, phenolate sodium, sodium benzenolate, sodium carbolate, phenol sodium, phenol sodium salt, phenol, sodium salt, naoph
PubChem CID 4445035
ChEBI CHEBI:52476
SMILES [Na+].[O-]C1=CC=CC=C1

Especificaciones

Melting Point >300°C
Odor Phenol-like
Linear Formula C6H5ONa
Quantity 50 g
UN Number UN2905
Sensitivity Air Sensitive
Solubility Information Very soluble in water,soluble in alcoholSoluble in water,acetone and alcohol.
IUPAC Name sodium;phenoxide
Formula Weight 116.09
Percent Purity 98%
Chemical Name or Material Sodium phenoxide
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Sodium phenoxide
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contain: Sodium phenolate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.