accessibility menu, dialog, popup

UserName

OHFFFF

CAS RN 141483-15-0

IUPAC Name: 2-fluoro-5-(trifluoromethyl)phenol
Synonyms: 2-fluoro-5-(trifluoromethyl)phenol
Molecular Weight (g/mol): 180.1
Molecular Formula: C7H4F4O
InChi Key: MCOSBFKOUQAIJS-UHFFFAOYSA-N
SMILES: OC1=CC(=CC=C1F)C(F)(F)F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)