missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)phenol, 98%, Thermo Scientific™

Product Code. 10796495 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10796495 1 g Glass bottle 1g
10420924 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10796495 Supplier Thermo Scientific Acros Supplier No. 397840010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 141483-15-0
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD00061286
InChI Key MCOSBFKOUQAIJS-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl phenol, 4-fluoro-3-hydroxybenzotrifluoride, 2-fluoro-5-trifluoromethyl-phenol, phenol, 2-fluoro-5-trifluoromethyl, alpha,alpha,alpha,6-tetrafluoro-m-cresol, pubchem4145, intermediates-zcf02079, acmc-1c6g6, ksc495m9p, 5-trifluoromethyl-2-fluorophenol
PubChem CID 518888
IUPAC Name 2-fluoro-5-(trifluoromethyl)phenol
SMILES OC1=CC(=CC=C1F)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Orange
Density 1.4360g/mL
Boiling Point 146.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H3(OH)CF3
Refractive Index 1.4380 to 1.4400 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.436
Formula Weight 180.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluoro-5-(trifluoromethyl)phenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.