accessibility menu, dialog, popup

UserName

CAS RN 14167-59-0

H3CCH3

CAS RN 14167-59-0

IUPAC Name: tetratriacontane
Synonyms: N-tetratriacontane
Molecular Weight (g/mol): 478.93
Molecular Formula: C34H70
InChi Key: GWVDBZWVFGFBCN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results