accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 142-19-8

IUPAC Name: prop-2-en-1-yl heptanoate
Synonyms: allyl heptanoate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: SJWKGDGUQTWDRV-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)OCC=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results