CAS RN 142217-69-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Entecavir, TRC
CAS: 142217-69-4 Molecular Formula: C12 H15 N5 O3 Synonym: 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-∣6H-Purin-6-one, 2-amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, [1S-(1α,3α,4β)]-∣BMS 200475∣Baraclude∣Entecavir∣SQ 34676
2
3
4