Learn More
Tocris Bioscience™ Entecavir
GREENER_CHOICE
Potent and selective hepatitis B virus inhibitor
Brand:  Tocris Bioscience™ 6234/50
Encompass
Product Code. 15787256
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Entecavir is a potent and selective hepatitis B virus reverse transcriptase inhibitor (EC50 = 3 nM); guanine analog. Inhibits hepatitis B virus replication in HepG2.2.15 cells. Displays selectively over HCMV, HSV-1, VZV, HIV and influenza (EC50 values are 15, 32, 30-60, >10 and >80 μM respectively). Orally bioavailable.
Specifications
Specifications
2-Amino-1,9-dihydro-9-[(1 S,3 R,4 S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-6 H-purin-6-one
142217-69-4
277.28
Store at -20°C
50 mg
Potent and Selective Hepatitis B Virus Inhibitor
C12H15N5O3
BMS 200475, SQ 34676
RNA/DNA Polymerase Inhibitors