CAS RN 143-19-1
CAS RN 143-19-1
IUPAC Name:
sodium (9Z)-octadec-9-enoate
Synonyms:
sodium oleate
Molecular Weight (g/mol):
304.45
Molecular Formula:
C18H33NaO2
InChi Key:
BCKXLBQYZLBQEK-KVVVOXFISA-M
SMILES:
[Na+].CCCCCCCC\C=C/CCCCCCCC([O-])=O