CAS RN 143130-94-3
CAS RN 143130-94-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(Z)-Azoxystrobin, TRC
CAS: 143130-94-3 Molecular Formula: C22 H17 N3 O5 Molecular Weight (g/mol): 403.39 Synonym: Benzeneacetic acid, 2-[[6-(2-cyanophenoxy)-4-pyrimidinyl]oxy]-α-(methoxymethylene)-, methyl ester, (Z)-,Z-Azoxystrobin,Benzeneacetic acid, 2-[[6-(2-cyanophenoxy)-4-pyrimidinyl]oxy]-α-(methoxymethylene)-, methyl ester, (αZ)-,(Z)-Azoxystrobin IUPAC Name: methyl (Z)-2-[2-[6-(2-cyanophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoate SMILES: CO\C=C(/C(=O)OC)\c1ccccc1Oc2cc(Oc3ccccc3C#N)ncn2