missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Azoxystrobin, TRC

Product Code. 18073640
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18073640 2.5 mg 2.5mg
18033150 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18073640 Supplier Toronto Research Chemicals Supplier No. TRCA9651552.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143130-94-3
Molecular Formula C22 H17 N3 O5
Molecular Weight (g/mol) 403.39
Synonym Benzeneacetic acid, 2-[[6-(2-cyanophenoxy)-4-pyrimidinyl]oxy]-α-(methoxymethylene)-, methyl ester, (Z)-, Z-Azoxystrobin, Benzeneacetic acid, 2-[[6-(2-cyanophenoxy)-4-pyrimidinyl]oxy]-α-(methoxymethylene)-, methyl ester, (αZ)-, (Z)-Azoxystrobin
IUPAC Name methyl (Z)-2-[2-[6-(2-cyanophenoxy)pyrimidin-4-yl]oxyphenyl]-3-methoxyprop-2-enoate
SMILES CO\C=C(/C(=O)OC)\c1ccccc1Oc2cc(Oc3ccccc3C#N)ncn2

Specifications

Analyte or Component Names (Z)-Azoxystrobin
Shipping Condition Room Temperature
InChI Formula InChI=1S/C22H17N3O5/c1-27-13-17(22(26)28-2)16-8-4-6-10-19(16)30-21-11-20(24-14-25-21)29-18-9-5-3-7-15(18)12-23/h3-11,13-14H,1-2H3/b17-13-
Solution Type Neat
Industry Type Forensics
Quantity 2.5 mg
Formula Weight 403.1168
Percent Purity >95%
Chemical Name or Material (Z)-Azoxystrobin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.