accessibility menu, dialog, popup

UserName

CAS RN 14348-41-5

HOOOHBr

CAS RN 14348-41-5

IUPAC Name: 3-bromo-4-hydroxybenzoic acid
Synonyms: 3-bromo-4-hydroxybenzoic acid
Molecular Weight (g/mol): 217.02
Molecular Formula: C7H5BrO3
InChi Key: XMEQDAIDOBVHEK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(O)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results