missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzoic acid, 97%

Product Code. 11499257
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499257 50 g 50g
11489257 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499257 Supplier Thermo Scientific Alfa Aesar Supplier No. B21338.18
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 14348-41-5 | C7H5BrO3 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14348-41-5
Molecular Formula C7H5BrO3
Molecular Weight (g/mol) 217.02
MDL Number MFCD00017547
InChI Key XMEQDAIDOBVHEK-UHFFFAOYSA-N
Synonym 3-bromo-4-hydroxy-benzoic acid, benzoic acid, 3-bromo-4-hydroxy, acmc-20aix6, 3-bromo-4-hydroxybenzoicacid, 3-bromo-4-hydroxybenzoic acid, 3-bromo-4-hydroxybenzoicacidhydrate, 3-bromo-4-hydroxybenzoic acid 10g
PubChem CID 84368
IUPAC Name 3-bromo-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(Br)=C1

Specifications

Melting Point 176°C to 178°C
Quantity 50 g
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-hydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.