accessibility menu, dialog, popup

UserName

FFSClOO

CAS RN 145758-05-0

IUPAC Name: 3,4-difluorobenzene-1-sulfonyl chloride
Synonyms: 3,4-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 212.59
Molecular Formula: C6H3ClF2O2S
InChi Key: FSGLUBQENACWCC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1F)S(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)