missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzenesulfonyl chloride, 97%

Product Code. 11324785
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324785 5 g 5g
11314785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324785 Supplier Thermo Scientific Alfa Aesar Supplier No. L17816.06
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 145758-05-0 | C6H3ClF2O2S | 212.595 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145758-05-0
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00153120
InChI Key FSGLUBQENACWCC-UHFFFAOYSA-N
Synonym 3,4-difluorobenzene-1-sulfonyl chloride, 3,4-difluorobenzenesulphonyl chloride, 3,4-difluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-difluoro, 3,4-difluorophenyl chlorosulfone, 3,4-difluorobenzenesulfonylchloride, 3,4-difluorobenzene sulfonyl chloride, 3,4-difluorobenzenesulphonylchloride, pubchem1958, acmc-1byq8
PubChem CID 2736913
IUPAC Name 3,4-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)F)F

Specifications

Density 1.586
Boiling Point 105°C (15 mmHg)
Refractive Index 1.514
Quantity 5 g
UN Number UN3265
Beilstein 4802506
Sensitivity Moisture sensitive
Formula Weight 212.6
Percent Purity 97%
Chemical Name or Material 3,4-Difluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.