accessibility menu, dialog, popup

UserName

CH3OONClClNH2N

CAS RN 1458-18-0

IUPAC Name: methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
Synonyms: methyl 3-amino-5,6-dichloropyrazine-2-carboxylate
Molecular Weight (g/mol): 222.03
Molecular Formula: C6H5Cl2N3O2
InChi Key: USYMCUGEGUFUBI-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(Cl)=C(Cl)N=C1N
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)