CAS RN 145986-07-8
CAS RN 145986-07-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(2R-cis)-1-[2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]-2,4(1H,3H)-pyrimidinedione, TRC
CAS: 145986-07-8 Molecular Formula: C8 H10 N2 O4 S Molecular Weight (g/mol): 230.24 Synonym: 2,4(1H,3H)-Pyrimidinedione, 1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, (2R-cis)- (9CI),1,3-Oxathiolane, 2,4(1H,3H)-pyrimidinedione deriv.,1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidine-2,4(1H,3H)-dione,Lamivudine Imp. J (EP) IUPAC Name: 1-[(2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidine-2,4-dione SMILES: OC[C@@H]1O[C@@H](CS1)N2C=CC(=O)NC2=O
2
1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidine-2,4(1H,3H)-dione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.