accessibility menu, dialog, popup

UserName

S

CAS RN 14633-54-6

IUPAC Name: (cyclopropylsulfanyl)benzene
Synonyms: (cyclopropylsulfanyl)benzene
Molecular Weight (g/mol): 150.24
Molecular Formula: C9H10S
InChi Key: YIBKCPJOFAUAKY-UHFFFAOYSA-N
SMILES: C1CC1SC1=CC=CC=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)