missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyl phenyl sulfide, 98%, ACROS Organics™

Product Code. 10553571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10553571 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10553571 Supplier Thermo Scientific Acros Supplier No. 210740010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 14633-54-6
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H10S
MDL Number MFCD00009684
Quantity 1 g
Synonym cyclopropyl phenyl sulfide, cyclopropyl phenyl sulfane, cyclopropylsulfanyl benzene, benzene, cyclopropylthio, cyclopropylphenylsulfide, cyclopropyl phenyl sulphide, unii-72c99lp48p, cyclopropylsulfanyl-benzene, pubchem10736, cyclopropylphenyl sulfide
InChI Key YIBKCPJOFAUAKY-UHFFFAOYSA-N
SMILES C1CC1SC1=CC=CC=C1
IUPAC Name (cyclopropylsulfanyl)benzene
Molecular Weight (g/mol) 150.24
PubChem CID 123348
Formula Weight 150.24
Chemical Name or Material Cyclopropyl phenyl sulfide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.