accessibility menu, dialog, popup

UserName

CAS RN 1470-61-7

IUPAC Name: silver(1+) (diethylcarbamothioyl)sulfanide
Synonyms: silver(1+) diethyldithiocarbamate
Molecular Weight (g/mol): 256.13
Molecular Formula: C5H10AgNS2
InChi Key: NSVHDIYWJVLAGH-UHFFFAOYSA-M
SMILES: [Ag+].CCN(CC)C([S-])=S
Physical Form 
  • (6)
  • (1)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
Grade 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)