missing translation for 'onlineSavingsMsg'
Learn More

Silver diethyldithiocarbamate, ACS

Product Code. 11381998
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381998 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11381998 Supplier Thermo Scientific Alfa Aesar Supplier No. 011898.09
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol

Silver diethyldithiocarbamate cluster plays a useful role for the deposition of acanthite (Ag2S) thin films which is employed in photoelectrochemical applications. Silver diethyldithiocarbamate membrane is used to develop ion selective electrodes for nitrate ion. It is useful in the colorimetric determination of arsenic. It acts as a copper chelator, and is utilized to quantitate copper in biological materials by ESR. It also works as an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1470-61-7
Molecular Formula C5H10AgNS2
Molecular Weight (g/mol) 256.13
MDL Number MFCD00004929
InChI Key NSVHDIYWJVLAGH-UHFFFAOYSA-M
Synonym silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
PubChem CID 3034078
IUPAC Name silver;N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)[S-].[Ag+]

Specifications

Melting Point 172°C to 175°C
Color White
Physical Form Powder
Quantity 10 g
Odor Odorless
Sensitivity Light sensitive
Appearance Yellow to pale brown powder
Solubility Information It is soluble in water and pyridine.
Formula Weight 256.14
Grade ACS
Chemical Name or Material Silver diethyldithiocarbamate, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.