accessibility menu, dialog, popup

UserName

CH3SOOOH

CAS RN 14763-60-1

IUPAC Name: 4-methanesulfonylphenol
Synonyms: 4-methanesulfonylphenol
Molecular Weight (g/mol): 172.2
Molecular Formula: C7H8O3S
InChi Key: KECCFSZFXLAGJS-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results