missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenol, 95%, Thermo Scientific™

Product Code. 10576992 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10576992 2.5 g Glass bottle 2.5g
10227323 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10576992 Supplier Thermo Scientific Acros Supplier No. 424880025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14763-60-1
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00047806
InChI Key KECCFSZFXLAGJS-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenol, 4-methanesulfonylphenol, 4-methanesulfonyl-phenol, 4-hydroxyphenyl methyl sulfone, 4-methylsulphonylphenol, phenol, p-methylsulfonyl, phenol, 4-methylsulfonyl, p-hydroxyphenyl methyl sulfone, p-methylsulfonyl phenol, unii-f7bif7zqj9
PubChem CID 123350
IUPAC Name 4-methylsulfonylphenol
SMILES CS(=O)(=O)C1=CC=C(O)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 90.0°C to 95.0°C
Color Brown-Pink to Tan
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4SO2CH3
Quantity 2.5 g
Solubility Information Solubility in water: slight. Other solubilities: soluble in dichloromethane,methanol
Formula Weight 172.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-(Methylsulfonyl)phenol
Show More Show Less
Product Identifier
  • 4-(Methylsulfonyl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.