accessibility menu, dialog, popup

UserName

CAS RN 1480-64-4

IUPAC Name: 3-chloro-2-fluoropyridine
Synonyms: 3-chloro-2-fluoropyridine
Molecular Weight (g/mol): 131.53
Molecular Formula: C5H3ClFN
InChi Key: IHGMHTQDGNVKTA-UHFFFAOYSA-N
SMILES: FC1=NC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results