accessibility menu, dialog, popup

UserName

CAS RN 1485-70-7

ONH

CAS RN 1485-70-7

IUPAC Name: N-benzylbenzamide
Synonyms: N-benzylbenzamide
Molecular Weight (g/mol): 211.26
Molecular Formula: C14H13NO
InChi Key: LKQUCICFTHBFAL-UHFFFAOYSA-N
SMILES: O=C(NCC1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)