missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylbenzamide, 99%

Product Code. 10164350 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10164350 25 g Plastic bottle 25g
10068731 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10164350 Supplier Thermo Scientific Acros Supplier No. 191170250
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1485-70-7 | C14H13NO | 211.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1485-70-7
Molecular Formula C14H13NO
Molecular Weight (g/mol) 211.26
MDL Number MFCD00003070
InChI Key LKQUCICFTHBFAL-UHFFFAOYSA-N
Synonym benzamide, n-phenylmethyl, n-benzyl-benzamide, phenyl-n-benzylcarboxamide, benzoylbenzylamine, n∼1∼-benzylbenzamide, benzamide, n-benzyl, n-phenylmethyl benzamide, acmc-1bx9y, n-benzylbenzamide, lkqucicfthbfal-uhfffaoysa
PubChem CID 73878
IUPAC Name N-benzylbenzamide
SMILES O=C(NCC1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 104.0°C to 107.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CONHCH2C6H5
Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 211.26
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Benzylbenzamide, 99%
Show More Show Less
compliance-icons
Product Identifier
  • N-Benzylbenzamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.