accessibility menu, dialog, popup

UserName

CAS RN 1489-69-6

O

CAS RN 1489-69-6

IUPAC Name: cyclopropanecarbaldehyde
Synonyms: cyclopropanecarbaldehyde
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: JMYVMOUINOAAPA-UHFFFAOYSA-N
SMILES: O=CC1CC1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (3)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)