missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarboxaldehyde, 98%

Product Code. 11323736
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11323736

Brand: Thermo Scientific Alfa Aesar L14931.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1489-69-6 | C4H6O | 70.091 g/mol

Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropanecarboxaldehyde reacts with Grignard or organolithium reagents to give the expected secondary alcohols. These are susceptible to ring opening in the presence of HBr, providing stereoselective access to homoallylic bromides. It is also used in the synthesis of eta(2)-enonenickel complexes by reacting with [Ni(cod)(2)] (cod = 1,5-cyclooctadiene) and PBu(3) and to prepare a cyclopropylidene tetralone which underwent an enantioselective palladium-catalyzed [3+2] TMM cycloaddition providing a chiral spiro-fused cyclopentene.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1489-69-6
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00012261
InChI Key JMYVMOUINOAAPA-UHFFFAOYSA-N
Synonym cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde
PubChem CID 123114
IUPAC Name cyclopropanecarbaldehyde
SMILES C1CC1C=O

Specifications

Density 0.938
Boiling Point 98°C to 101°C
Flash Point 7°C (44°F)
Odor Stench
Refractive Index 1.4298
Quantity 5 g
UN Number UN2924
Beilstein 906781
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxaldehyde
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.