accessibility menu, dialog, popup

UserName

CAS RN 149739-65-1

IUPAC Name: 5-(3-bromophenyl)-1H-pyrazole
Synonyms: 3-(3-bromophenyl)-2H-pyrazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: NVRXIZHZQPRBKL-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CC=NN1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results