missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Bromophenyl)-1H-pyrazole, ≥97%, Thermo Scientific™

Product Code. 10269672
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10269672 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10269672 Supplier Thermo Scientific Maybridge Supplier No. CC31710DA
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 149739-65-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H7BrN2
MDL Number MFCD01940433
Quantity 1 g
Synonym 3-3-bromophenyl-1h-pyrazole, 5-3-bromophenyl-1h-pyrazole, 5-3-bromo-phenyl-1h-pyrazole, 3-3-bromophenyl pyrazole, 3-3-bromophenyl-2h-pyrazole, 1h-pyrazole, 3-3-bromophenyl, 1-bromo-3-1h-pyrazol-3-yl benzene, 5-3-bromophenyl pyrazole, acmc-1c2w8, ksc494i0r
InChI Key NVRXIZHZQPRBKL-UHFFFAOYSA-N
SMILES BrC1=CC=CC(=C1)C1=CC=NN1
IUPAC Name 5-(3-bromophenyl)-1H-pyrazole
Molecular Weight (g/mol) 223.07
PubChem CID 2735613
Formula Weight 223.07
Percent Purity 97%
Chemical Name or Material 3-(3-Bromophenyl)-1H-pyrazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.