accessibility menu, dialog, popup

UserName

H3CNCH3ONHCl

CAS RN 150-68-5

IUPAC Name: 1-(4-chlorophenyl)-3,3-dimethylurea
Synonyms: monuron karmex monuron herbicide caswell no. 583 monuuron epa pesticide chemical code 035501 monuron herbicides telvar telvar W. monuron weedkiller chlorfenidim CMU
Molecular Weight (g/mol): 198.65
Molecular Formula: C9H11ClN2O
InChi Key: BMLIZLVNXIYGCK-UHFFFAOYSA-N
SMILES: CN(C)C(=O)NC1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results