accessibility menu, dialog, popup

UserName

CH3BrF

CAS RN 151412-12-3

IUPAC Name: 1-(bromomethyl)-2-fluoro-3-methylbenzene
Synonyms: 1-(bromomethyl)-2-fluoro-3-methylbenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: DCGWFNZSJBIAKE-UHFFFAOYSA-N
SMILES: CC1=C(F)C(CBr)=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results