missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)-2-fluoro-3-methylbenzene, ≥95%, Maybridge

Product Code. 10644395
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10644395 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10644395 Supplier Thermo Scientific Maybridge Supplier No. CD05588DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 151412-12-3
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C8H8BrF
MDL Number MFCD00042470
Quantity 1 g
Synonym 2-fluoro-3-methylbenzyl bromide, 1-bromomethyl-2-fluoro-3-methylbenzene, benzene,1-bromomethyl-2-fluoro-3-methyl, acmc-20anvi, 2-fluoro-3-methylbenzylbromide, 2-fluoro-3-methyl-benzylbromide, 2-fluoro-3-methyl benzyl bromide, 3-bromomethyl-2-fluoro-1-methylbenzene, 1-bromomethyl-2-fluoro-3-methylbenzene #
InChI Key DCGWFNZSJBIAKE-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)CBr)F
IUPAC Name 1-(bromomethyl)-2-fluoro-3-methylbenzene
Molecular Weight (g/mol) 203.054
PubChem CID 519062
Formula Weight 203.05
Percent Purity 95%
Chemical Name or Material 1-(Bromomethyl)-2-fluoro-3-methylbenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.