accessibility menu, dialog, popup

UserName

CAS RN 15184-99-3

CH3NH3COCH3

CAS RN 15184-99-3

IUPAC Name: [(3-methoxyphenyl)methyl]dimethylamine
Synonyms: [(3-methoxyphenyl)methyl]dimethylamine
Molecular Weight (g/mol): 165.24
Molecular Formula: C10H15NO
InChi Key: IENUHHADGQLQLU-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CN(C)C)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results