missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-N,N-dimethylbenzylamine, 98%

Product Code. 15491448
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491448 5 g 5g
15481448 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491448 Supplier Thermo Scientific Alfa Aesar Supplier No. H55260.06
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15184-99-3 | C10H15NO | 165.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15184-99-3
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.236
MDL Number MFCD00011530
InChI Key IENUHHADGQLQLU-UHFFFAOYSA-N
Synonym 3-methoxy-n,n-dimethylbenzylamine, 3-methoxyphenyl methyl dimethylamine, 3-methoxybenzyl dimethylamine, 1-3-methoxyphenyl-n,n-dimethylmethanamine, benzenemethanamine, 3-methoxy-n,n-dimethyl, acmc-20an1l, n,n-dimethyl-3-methoxybenzylamine, 3-methoxy-n,n-dimethyl benzylamine, 3-methoxy-n,n-dimethyl-benzenemethanamine
PubChem CID 547434
IUPAC Name 1-(3-methoxyphenyl)-N,N-dimethylmethanamine
SMILES CN(C)CC1=CC(=CC=C1)OC

Specifications

Color Colorless
Density 0.984 g/mL
Boiling Point 105°C (13 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.514
Quantity 5 g
Formula Weight 165.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Methoxy-N,N-dimethylbenzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.