accessibility menu, dialog, popup

UserName

CAS RN 15219-34-8

BrOBrO

CAS RN 15219-34-8

IUPAC Name: oxalic dibromide
Synonyms: oxalic dibromide
Molecular Weight (g/mol): 215.83
Molecular Formula: C2Br2O2
InChi Key: JAZLVNXWYDFQFE-UHFFFAOYSA-N
SMILES: BrC(=O)C(Br)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)