missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl bromide, 98%

Product Code. 10662762 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662762 100 g Glass bottle 100g
10112291 25 g Glass bottle 25g
10155773 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10662762 Supplier Thermo Scientific Acros Supplier No. 150891000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 15219-34-8 | C2Br2O2 | 215.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 15219-34-8
Molecular Formula C2Br2O2
Molecular Weight (g/mol) 215.83
MDL Number MFCD00000113
InChI Key JAZLVNXWYDFQFE-UHFFFAOYSA-N
Synonym oxalyl bromide, ethanedioyl dibromide, ethanedioyl bromide, oxalic dibromide, brcocobr, oxalyl bromide solution, ethanedioyl dibromide solution, oxalyl bromide solution, 2.0 m in methylene chloride
PubChem CID 84841
IUPAC Name oxalyl dibromide
SMILES BrC(=O)C(Br)=O

Spezifikation

Melting Point -19.0°C
Density 2.3000g/mL
Boiling Point 102.0°C to 103.0°C (720.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrCOCOBr
Refractive Index 1.5205 to 1.5225
Quantity 100 g
Fieser 01,767; 01,1137
Specific Gravity 2.3
Solubility Information Solubility in water: reacts with water.
Formula Weight 215.83
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Oxalyl bromide
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.