accessibility menu, dialog, popup

UserName

CAS RN 15244-10-7

FeOSOOOOSOOOOSOOOFe

CAS RN 15244-10-7

IUPAC Name: diiron(3+) hydrate trisulfate
Synonyms: diiron(3+) hydrate trisulfate
Molecular Weight (g/mol): 417.87
Molecular Formula: Fe2H2O13S3
InChi Key: VXWSFRMTBJZULV-UHFFFAOYSA-H
SMILES: O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Grade 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)